| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700038963.mm.1 |
NM_001252572.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700038963.mm.1 |
NM_001252572.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700038963.mm.1 |
NM_007803.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700038963.mm.1 |
NM_007803.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700038963.mm.1 |
TC0700004604.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700038961.mm.1 |
NM_007803.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700038961.mm.1 |
NM_007803.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700038961.mm.1 |
TC0700004604.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700038962.mm.1 |
NM_001252572.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700038962.mm.1 |
NM_001252572.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700038962.mm.1 |
NM_007803.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700038962.mm.1 |
NM_007803.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700038962.mm.1 |
TC0700004604.mm.1 |
MTA TC |