Assay Details
| Gene Symbol: Cttn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007803.5 | NP_031829.2 | 11 - 12 | 1241 | 65 | |
| GenBank mRNA | AK146856.1 | - | 11 - 12 | 1048 | 65 | |
| AK148010.1 | - | 11 - 12 | 1044 | 65 | ||
| AK168699.1 | - | 11 - 12 | 1042 | 65 | ||
| BC117820.1 | - | 10 - 11 | 943 | 65 | ||
| U03184.1 | - | 11 - 12 | 65 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Cttn
Genomic Map