Assay Details
| Gene Symbol: Cttn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252572.1 | NP_001239501.1 | 12 - 13 | 1254 | 99 | |
| NM_007803.5 | NP_031829.2 | 13 - 14 | 1365 | 99 | ||
| GenBank mRNA | AK084249.1 | - | 12 - 13 | 1064 | 99 | |
| AK146856.1 | - | 13 - 14 | 1172 | 99 | ||
| AK165555.1 | - | 12 - 13 | 1057 | 99 | ||
| AK168699.1 | - | 13 - 14 | 1166 | 99 | ||
| AK168871.1 | - | 12 - 13 | 1055 | 99 | ||
| AK168945.1 | - | 12 - 13 | 1065 | 99 | ||
| AK197563.1 | - | 2 - 3 | 104 | 99 | ||
| BC011434.1 | - | 12 - 13 | 1047 | 3981559 | 99 | |
| BC117820.1 | - | 12 - 13 | 1067 | 99 | ||
| U03184.1 | - | 13 - 14 | 1154 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cttn
Genomic Map