Assay Details
| Gene Symbol: Dnm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039520.2 | NP_001034609.1 | 15 - 16 | 1856 | 75 | |
| NM_001253893.1 | NP_001240822.1 | 15 - 16 | 1856 | 75 | ||
| NM_001253894.1 | NP_001240823.1 | 15 - 16 | 1856 | 75 | ||
| NM_007871.2 | NP_031897.2 | 14 - 15 | 1844 | 75 | ||
| GenBank mRNA | AK005012.2 | - | 15 - 16 | 1832 | 75 | |
| AK150046.1 | - | 15 - 16 | 1819 | 75 | ||
| AK154375.1 | - | 13 - 14 | 1508 | 75 | ||
| AK170574.1 | - | 15 - 16 | 1832 | 75 | ||
| AK171049.1 | - | 15 - 16 | 1810 | 75 | ||
| AK171566.1 | - | 15 - 16 | 1820 | 75 | ||
| AK172231.1 | - | 15 - 16 | 1837 | 75 | ||
| BC010233.1 | - | 6 - 7 | 572 | 3499181 | 75 | |
| L31398.1 | - | 14 - 15 | 1784 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dnm2
Genomic Map