Assay Details
| Gene Symbol: Dtnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162465.1 | NP_001155937.1 | 6 - 7 | 858 | 78 | |
| NM_007886.2 | NP_031912.2 | 6 - 7 | 793 | 78 | ||
| NR_027869.1 | 6 - 7 | 858 | 78 | |||
| NR_027870.1 | 6 - 7 | 858 | 78 | |||
| GenBank mRNA | AJ003007.1 | - | 6 - 7 | 726 | 78 | |
| AK043436.1 | - | 6 - 7 | 816 | 78 | ||
| AK083752.1 | - | 6 - 7 | 814 | 78 | ||
| AK085675.1 | - | 6 - 7 | 784 | 78 | ||
| BC030924.1 | - | 6 - 7 | 793 | 4910166 | 78 | |
| Y15742.1 | - | 6 - 7 | 690 | 78 | ||
More Information
Set Membership: |
|

Dtnb
Genomic Map