Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300019251.mm.1 |
NM_001252653.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300019251.mm.1 |
NM_007899.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300019251.mm.1 |
TC0300002482.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300019249.mm.1 |
NM_001252653.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300019249.mm.1 |
NM_007899.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300019249.mm.1 |
TC0300002482.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300019253.mm.1 |
NM_001252653.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300019253.mm.1 |
NM_007899.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300019253.mm.1 |
TC0300002482.mm.1 |
MTA TC |