Assay Details
| Gene Symbol: Eps8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271587.1 | NP_001258516.1 | 18 - 19 | 2367 | 70 | |
| NM_001271588.1 | NP_001258517.1 | 18 - 19 | 2298 | 70 | ||
| NM_001271589.1 | NP_001258518.1 | 11 - 12 | 1368 | 70 | ||
| NM_001271595.1 | NP_001258524.1 | 18 - 19 | 2372 | 70 | ||
| NM_007945.3 | NP_031971.2 | 19 - 20 | 2373 | 70 | ||
| GenBank mRNA | AK136246.1 | - | 9 - 10 | 1140 | 70 | |
| AK149683.1 | - | 18 - 19 | 2161 | 70 | ||
| AK155022.1 | - | 18 - 19 | 2133 | 70 | ||
| AK165684.1 | - | 11 - 12 | 1371 | 70 | ||
| AK166156.1 | - | 19 - 20 | 2208 | 70 | ||
| BC016890.1 | - | 18 - 19 | 2384 | 4240899 | 70 | |
| L21671.1 | - | 18 - 19 | 2288 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Eps8
Genomic Map