Assay Details
| Gene Symbol: Fst | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008046.3 | NP_032072.1 | 4 - 5 | 1084 | 74 | |
| GenBank mRNA | Z29532.1 | - | 4 - 5 | 719 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fst
Genomic Map