Assay Details
| Gene Symbol: Fst | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301373.1 | NP_001288302.1 | 5 - 6 | 1310 | 144 | |
| NM_008046.3 | NP_032072.1 | 5 - 6 | 1307 | 144 | ||
| GenBank mRNA | BC144926.1 | - | 5 - 6 | 1134 | 9053474 | 144 |
| BC145945.1 | - | 5 - 6 | 1134 | 40131248 | 144 | |
| Z29532.1 | - | 5 - 6 | 942 | 144 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fst
Genomic Map