Assay Details
| Gene Symbol: Gclm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008129.4 | NP_032155.1 | 2 - 3 | 617 | 76 | |
| GenBank mRNA | AK028236.1 | - | 2 - 3 | 583 | 76 | |
| AK172387.1 | - | 2 - 3 | 558 | 76 | ||
| AK176933.1 | - | 2 - 3 | 76 | |||
| BC132404.1 | - | 3 - 4 | 304 | 40130681 | 76 | |
| BC138783.1 | - | 3 - 4 | 326 | 8861805 | 76 | |
| CT010256.1 | - | 2 - 3 | 188 | 76 | ||
| U95053.1 | - | 2 - 3 | 228 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gclm
Genomic Map