Assay Details
| Gene Symbol: Gnb2l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008143.3 | NP_032169.1 | 5 - 6 | 745 | 65 | |
| GenBank mRNA | AK002493.1 | - | 5 - 6 | 734 | 65 | |
| AK012242.1 | - | 4 - 5 | 532 | 65 | ||
| AK017772.1 | - | 5 - 6 | 746 | 65 | ||
| AK051226.1 | - | 5 - 6 | 746 | 65 | ||
| AK159797.1 | - | 5 - 6 | 746 | 65 | ||
| AK160528.1 | - | 5 - 6 | 716 | 65 | ||
| AK160739.1 | - | 5 - 6 | 743 | 65 | ||
| AK161234.1 | - | 5 - 6 | 746 | 65 | ||
| AK166417.1 | - | 5 - 6 | 734 | 65 | ||
| AK166786.1 | - | 5 - 6 | 748 | 65 | ||
| AK166800.1 | - | 5 - 6 | 749 | 65 | ||
| AK166868.1 | - | 1 - 2 | 3644 | 65 | ||
| AK166889.1 | - | 4 - 5 | 386 | 65 | ||
| AK166945.1 | - | 5 - 6 | 746 | 65 | ||
| AK168086.1 | - | 5 - 6 | 746 | 65 | ||
| AK168096.1 | - | 5 - 6 | 746 | 65 | ||
| AK168196.1 | - | 5 - 6 | 746 | 65 | ||
| AK168349.1 | - | 5 - 6 | 777 | 65 | ||
| AK169612.1 | - | 5 - 6 | 745 | 65 | ||
| BC020322.1 | - | 5 - 6 | 722 | 65 | ||
| BC036295.1 | - | 3 - 4 | 285 | 65 | ||
| BC046760.1 | - | 5 - 6 | 730 | 5693613 | 65 | |
| D29802.1 | - | 5 - 6 | 739 | 65 | ||
| X75313.1 | - | 5 - 6 | 745 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gnb2l1
Genomic Map