Assay Details
| Gene Symbol: Gpc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008150.2 | NP_032176.2 | 4 - 5 | 1029 | 71 | |
| GenBank mRNA | BC006622.1 | - | 4 - 5 | 1029 | 3967797 | 71 |
| CF168390.1 | - | 1 - 2 | 52 | 71 | ||
| CN710713.1 | - | 2 - 3 | 507 | 71 | ||
| X83577.1 | - | 4 - 5 | 1350 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gpc4
Genomic Map