Assay Details
| Gene Symbol: Gss | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291111.1 | NP_001278040.1 | 2 - 3 | 262 | 101 | |
| NM_008180.2 | NP_032206.1 | 3 - 4 | 397 | 101 | ||
| GenBank mRNA | AK146486.1 | - | 3 - 4 | 302 | 101 | |
| AK149577.1 | - | 2 - 3 | 167 | 101 | ||
| AK159630.1 | - | 3 - 4 | 322 | 101 | ||
| AK169096.1 | - | 3 - 4 | 317 | 101 | ||
| AK212683.1 | - | 1 - 2 | 139 | 101 | ||
| U35456.1 | - | 2 - 3 | 293 | 101 | ||
More Information
Set Membership: |
|

Gss
Genomic Map