Assay Details
| Gene Symbol: Gss | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291111.1 | NP_001278040.1 | 9 - 10 | 1021 | 67 | |
| NM_008180.2 | NP_032206.1 | 10 - 11 | 1156 | 67 | ||
| GenBank mRNA | AK146486.1 | - | 10 - 11 | 1061 | 67 | |
| AK149577.1 | - | 9 - 10 | 926 | 67 | ||
| AK159630.1 | - | 10 - 11 | 1081 | 67 | ||
| AK169096.1 | - | 10 - 11 | 1076 | 67 | ||
| BC003784.1 | - | 10 - 11 | 1069 | 3593913 | 67 | |
| U35456.1 | - | 9 - 10 | 1052 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gss
Genomic Map