Assay Details
| Gene Symbol: Hpgd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008278.2 | NP_032304.2 | 5 - 6 | 566 | 60 | |
| GenBank mRNA | AK002472.1 | - | 5 - 6 | 532 | 60 | |
| AK146038.1 | - | 5 - 6 | 534 | 60 | ||
| BC021157.1 | - | 5 - 6 | 525 | 4208980 | 60 | |
| U44389.1 | - | 5 - 6 | 525 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hpgd
Genomic Map