Assay Details
| Gene Symbol: Idua | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008325.4 | NP_032351.2 | 9 - 10 | 1607 | 85 | |
| NM_172997.1 | NP_766585.1 | 8 - 9 | 1466 | 85 | ||
| GenBank mRNA | AK031414.1 | - | 8 - 9 | 1261 | 85 | |
| AK045330.1 | - | 7 - 8 | 1094 | 85 | ||
| AK155627.1 | - | 9 - 10 | 1608 | 85 | ||
| AK169998.1 | - | 8 - 9 | 1265 | 85 | ||
| AK171124.1 | - | 4 - 5 | 618 | 85 | ||
| BC057935.1 | - | 8 - 9 | 1228 | 5327150 | 85 | |
| L34111.1 | - | 8 - 9 | 1181 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Idua
Genomic Map