Assay Details
| Gene Symbol: Ifit1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008331.3 | NP_032357.2 | 1 - 2 | 97 | 80 | |
| GenBank mRNA | AK089608.1 | - | 1 - 2 | 104 | 80 | |
| AK131669.1 | - | 1 - 2 | 94 | 80 | ||
| AK144230.1 | - | 1 - 2 | 102 | 80 | ||
| AK150352.1 | - | 1 - 2 | 104 | 80 | ||
| AK150424.1 | - | 1 - 2 | 98 | 80 | ||
| AK150609.1 | - | 1 - 2 | 107 | 80 | ||
| AK151125.1 | - | 1 - 2 | 107 | 80 | ||
| AK151158.1 | - | 1 - 2 | 98 | 80 | ||
| AK151278.1 | - | 1 - 2 | 107 | 80 | ||
| AK151298.1 | - | 1 - 2 | 101 | 80 | ||
| AK151983.1 | - | 1 - 2 | 100 | 80 | ||
| AK152112.1 | - | 1 - 2 | 100 | 80 | ||
| AK152753.1 | - | 1 - 2 | 98 | 80 | ||
| AK153184.1 | - | 1 - 2 | 100 | 80 | ||
| AK153500.1 | - | 1 - 2 | 98 | 80 | ||
| AK153532.1 | - | 1 - 2 | 107 | 80 | ||
| AK157848.1 | - | 1 - 2 | 98 | 80 | ||
| BC003768.1 | - | 1 - 2 | 81 | 3496673 | 80 | |
| U43084.1 | - | 1 - 2 | 72 | 80 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK089608.1,AK144230.1,AK150352.1,AK150424.1,AK150609.1,AK151125.1,AK151158.1,AK151278.1,AK151298.1,AK152753.1,AK153184.1,AK153500.1,AK153532.1,AK157848.1,BC003768.1,NM_008331.3,U43084.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P327751 | U43084.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P327751 | AK144230.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P327751 | U43084.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P327751 | AK144230.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 790323 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 750331 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 614822 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 485355 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 306632 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 130435 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P100920 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P327751 | U43084.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P327751 | AK144230.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P327751 | U43084.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P327751 | AK144230.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32081355 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32081358 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32081364 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32081354 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32081365 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32081359 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32081356 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32081362 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32081361 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32081368 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32081366 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09158620 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P100920 | - | - |
| Whole Mouse Genome G4122A | A_51_P327751 | U43084.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P327751 | AK144230.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P327751 | U43084.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P327751 | AK144230.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111203 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P120242 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P100920 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14249587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081361 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14334318 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32081457 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32081365 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32081366 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32081360 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32081364 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32081362 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32081379 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14248115 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32081393 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32081376 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32081462 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32081371 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32081355 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32081388 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32081390 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32081368 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32081445 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32081383 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14385368 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14255872 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14449210 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14339076 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32081395 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32081382 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14255863 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32081402 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14339044 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32081396 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14338917 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14339081 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14195165 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14449217 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32081377 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32081456 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32081358 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32081450 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32081386 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32081359 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32081463 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32081387 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32081448 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32081451 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32081454 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32081459 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32081458 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32081356 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10462626 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10462627 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10462624 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10462625 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1450783_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10462623 | - | - |
| Mouse Genome 430 2.0 Array | 1450783_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1450783_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 15957_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 15957_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1900004480.mm.1 | NM_008331.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900004480.mm.1 | NM_008331.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900004480.mm.1 | TC1900000504.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900004477.mm.1 | NM_008331.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900004477.mm.1 | NM_008331.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900004477.mm.1 | TC1900000504.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900004479.mm.1 | NM_008331.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900004479.mm.1 | NM_008331.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900004479.mm.1 | TC1900000504.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900004478.mm.1 | NM_008331.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900004478.mm.1 | NM_008331.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900004478.mm.1 | TC1900000504.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1450783_at | - | - |
| Mouse Expression Set 430 | 1450783_PM_at | - | - |
| Murine Genome U74v2 Set | 100981_PM_at | - | - |
| Murine Genome U74v2 Set | 100981_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450783_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450783_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450783_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450783_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 607896 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5270373 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5270373 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005270373 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2774346 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2774346 | - | - |
| mouse-6 v1.1 (Illumina) | scl53191.3_226-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5270373 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl53191.3_226-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5270373 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2774340 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2774340 | - | - |
Citations (1)
Distinct RIG-I and MDA5 signaling by RNA viruses in innate immunity.
- Authors
- Loo YM, Fornek J, Crochet N, Bajwa G, Perwitasari O, Martinez-Sobrido L, Akira S, Gill MA, Garcia-Sastre A, Katze MG, Gale M Jr
- Journal
- J Virol - Page No 335 (Tue Jan 01 00:00:00 EST 2008)
- Volume (Year)
- 82 (2008)
- Product Usage
- -

Ifit1
Genomic Map