Assay Details
| Gene Symbol: Igfbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008343.2 | NP_032369.2 | 2 - 3 | 770 | 74 | |
| GenBank mRNA | AK077477.1 | - | 2 - 3 | 311 | 74 | |
| AK141690.1 | - | 2 - 3 | 392 | 74 | ||
| BC031421.1 | - | 2 - 3 | 729 | 74 | ||
| BC058261.1 | - | 2 - 3 | 740 | 6487655 | 74 | |
| X81581.1 | - | 2 - 3 | 718 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Igfbp3
Genomic Map