Assay Details
| Gene Symbol: Kif2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145779.1 | NP_001139251.1 | 15 - 16 | 1608 | 72 | |
| NM_008442.2 | NP_032468.2 | 16 - 17 | 1991 | 72 | ||
| GenBank mRNA | AK037037.1 | - | 15 - 16 | 1521 | 72 | |
| BC006803.1 | - | 15 - 16 | 1603 | 3152883 | 72 | |
| BE863830.1 | - | 2 - 3 | 164 | 72 | ||
| D12644.1 | - | 16 - 17 | 1992 | 72 | ||
| Y15894.1 | - | 14 - 15 | 1458 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kif2a
Genomic Map