Assay Details
| Gene Symbol: Kif5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008448.3 | NP_032474.2 | 11 - 12 | 1531 | 76 | |
| GenBank mRNA | BC090841.1 | - | 11 - 12 | 1507 | 30543821 | 76 |
| L27153.1 | - | 11 - 12 | 1125 | 76 | ||
| L29223.2 | - | 1 - 2 | 39 | 76 | ||
| U86090.1 | - | 11 - 12 | 1358 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kif5b
Genomic Map