Assay Details
| Gene Symbol: Dpysl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009468.5 | NP_033494.1 | 1 - 2 | 387 | 67 | |
| GenBank mRNA | BC023003.1 | - | 1 - 2 | 71 | 5367381 | 67 |
| X87817.1 | - | 1 - 2 | 347 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dpysl3
Genomic Map