Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900012689.mm.1 |
NM_001200002.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900012689.mm.1 |
NM_001200002.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900012689.mm.1 |
NM_009752.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900012689.mm.1 |
NR_108101.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900012689.mm.1 |
NR_108102.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900012689.mm.1 |
TC0900001513.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900012694.mm.1 |
NM_009752.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900012694.mm.1 |
TC0900001513.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900012688.mm.1 |
NM_001200002.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900012688.mm.1 |
NM_001200002.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900012688.mm.1 |
NM_009752.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900012688.mm.1 |
NR_108101.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900012688.mm.1 |
NR_108102.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900012688.mm.1 |
TC0900001513.mm.1 |
MTA TC |