Assay Details
| Gene Symbol: Cd19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009844.2 | NP_033974.2 | 3 - 4 | 587 | 77 | |
| GenBank mRNA | AK089835.1 | - | 3 - 4 | 586 | 77 | |
| AK156125.1 | - | 3 - 4 | 590 | 77 | ||
| AK163608.1 | - | 1 - 2 | 1084 | 77 | ||
| AK172008.1 | - | 3 - 4 | 549 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cd19
Genomic Map