Assay Details
| Gene Symbol: Cd22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043317.2 | NP_001036782.1 | 7 - 8 | 1412 | 99 | |
| NM_009845.3 | NP_033975.3 | 5 - 6 | 1287 | 99 | ||
| NR_102722.1 | 7 - 8 | 1484 | 99 | |||
| GenBank mRNA | AF102134.1 | - | 4 - 5 | 1056 | 99 | |
| AF115400.1 | - | 4 - 5 | 1056 | 99 | ||
| AF115401.1 | - | 4 - 5 | 1056 | 99 | ||
| AK041230.1 | - | 7 - 8 | 1419 | 99 | ||
| AK143835.1 | - | 7 - 8 | 1350 | 99 | ||
| AK156735.1 | - | 5 - 6 | 1286 | 99 | ||
| AK171589.1 | - | 4 - 5 | 1093 | 99 | ||
| AK171990.1 | - | 4 - 5 | 1407 | 99 | ||
| AK172301.1 | - | 5 - 6 | 1243 | 99 | ||
| BC150807.1 | - | 6 - 7 | 1371 | 9087718 | 99 | |
| L02844.1 | - | 6 - 7 | 1558 | 99 | ||
| L16928.1 | - | 6 - 7 | 1535 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cd22
Genomic Map