Assay Details
| Gene Symbol: Ch25h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009890.1 | NP_034020.1 | 1 | 845 | 92 | |
| GenBank mRNA | AF059213.1 | - | 1 | 845 | 92 | |
| AK140360.1 | - | 1 | 863 | 92 | ||
| AK152770.1 | - | 1 | 854 | 92 | ||
| AK160657.1 | - | 1 | 859 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ch25h
Genomic Map