Assay Details
| Gene Symbol: Cish | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009895.3 | NP_034025.1 | 1 - 2 | 351 | 81 | |
| GenBank mRNA | AK077338.1 | - | 1 - 2 | 351 | 81 | |
| AK170554.1 | - | 1 - 2 | 152 | 81 | ||
| AK170877.1 | - | 1 - 2 | 147 | 81 | ||
| AK171431.1 | - | 1 - 2 | 152 | 81 | ||
| BC003783.1 | - | 1 - 2 | 155 | 3487580 | 81 | |
| BC022178.1 | - | 1 - 2 | 116 | 4193243 | 81 | |
| D31943.1 | - | 1 - 2 | 187 | 81 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK077338.1,AK170554.1,AK170877.1,AK171431.1,BC003783.1,BC022178.1,D31943.1,NM_009895.3 transcripts.
Set Membership: |
|

Cish
Genomic Map