Assay Details
| Gene Symbol: Cldn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009902.4 | NP_034032.1 | 1 | 1127 | 60 | |
| GenBank mRNA | AF095905.1 | - | 1 | 951 | 60 | |
| AK002672.1 | - | 1 | 1128 | 60 | ||
| AK006514.1 | - | 1 | 1089 | 60 | ||
| AK012677.1 | - | 1 | 1079 | 60 | ||
| AK165808.1 | - | 1 | 771 | 60 | ||
| BC012650.1 | - | 3 | 789 | 4217040 | 60 | |
| BC023269.1 | - | 1 | 803 | 60 | ||
| BC032070.1 | - | 1 | 495 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24114167 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24114171 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24114169 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24114166 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24114170 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24114173 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24114164 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24114172 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2183473 | AK002672.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P118393 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2183473 | AK002672.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P157902 | BC012650.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P157902 | BC023269.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P157902 | BC032070.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P157902 | AK002672.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P157902 | AK006514.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114162 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114156 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114170 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114159 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114163 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114177 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114174 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114169 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114145 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114155 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114164 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114143 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114175 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114148 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114165 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114167 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114181 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114172 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114179 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114150 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114176 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114149 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114178 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24114153 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P118393 | - | - |
| Whole Mouse Genome G4122A | A_51_P157902 | BC012650.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P157902 | BC023269.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P157902 | BC032070.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P157902 | AK002672.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P157902 | AK006514.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108361 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P118393 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04664696 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04664688 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04664704 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04664692 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04664685 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04664693 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04664676 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24114171 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24114167 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24114166 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24114172 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24114170 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24114173 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24114169 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24114164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24114188 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04664703 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04660405 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04664685 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24114145 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24114159 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04164630 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24114204 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24114165 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04164629 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24114185 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24114174 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24114190 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04664696 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24114162 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04970163 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04535212 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24114200 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24114156 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04327614 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04970164 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04512195 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04987302 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24114176 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24114181 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24114205 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04164632 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24114149 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24114147 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24114154 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24114138 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24114182 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24114207 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04968382 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24114191 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24114150 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04297062 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24114167 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24114135 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04970168 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04968379 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04209471 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04968381 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24114153 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24114213 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24114170 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04579525 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04934382 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24114155 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24114143 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24114179 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24114211 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24114134 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04664678 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24114164 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24114208 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24114175 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10526245 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526248 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526247 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526243 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526244 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526249 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526246 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526242 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1460569_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1434651_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1451701_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1426332_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10526241 | - | - |
| Mouse Genome 430 2.0 Array | 1453258_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1426332_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1434651_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1434651_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1426332_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1460569_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1451701_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1451701_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1460569_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1453258_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12739_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12739_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500014031.mm.1 | TC0500001570.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500014031.mm.1 | NM_009902.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500014031.mm.1 | NM_009902.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500014033.mm.1 | TC0500001570.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500014033.mm.1 | NM_009902.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500014033.mm.1 | NM_009902.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500014034.mm.1 | TC0500001570.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500014034.mm.1 | NM_009902.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500014034.mm.1 | NM_009902.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500014032.mm.1 | TC0500001570.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500014032.mm.1 | NM_009902.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500014032.mm.1 | NM_009902.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500014030.mm.1 | TC0500001570.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500014030.mm.1 | NM_009902.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500014030.mm.1 | NM_009902.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500014035.mm.1 | TC0500001570.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500014035.mm.1 | NM_009902.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500014035.mm.1 | NM_009902.4 | Refseq |
| Mouse Expression Set 430 | 1426332_PM_a_at | - | - |
| Mouse Expression Set 430 | 1451701_x_at | - | - |
| Mouse Expression Set 430 | 1451701_PM_x_at | - | - |
| Mouse Expression Set 430 | 1453258_PM_at | - | - |
| Mouse Expression Set 430 | 1460569_PM_x_at | - | - |
| Mouse Expression Set 430 | 1460569_x_at | - | - |
| Mouse Expression Set 430 | 1434651_PM_a_at | - | - |
| Mouse Expression Set 430 | 1434651_a_at | - | - |
| Mouse Expression Set 430 | 1453258_at | - | - |
| Mouse Expression Set 430 | 1426332_a_at | - | - |
| Murine Genome U74v2 Set | 94493_PM_at | - | - |
| Murine Genome U74v2 Set | 162315_PM_f_at | - | - |
| Murine Genome U74v2 Set | 94493_at | - | - |
| Murine Genome U74v2 Set | 162315_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1434651_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451701_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426332_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1434651_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1453258_at | - | - |
| Mouse Genome 430A 2.0 Array | 1460569_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1453258_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451701_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1460569_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426332_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426332_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426332_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451701_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1453258_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1460569_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1434651_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1434651_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1460569_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451701_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1453258_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 929248 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2570524 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2570524 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002570524 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2634167 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2634167 | - | - |
| mouse-6 v1.1 (Illumina) | scl27140.1_204-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2570524 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl27140.1_204-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2570524 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2634167 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2634167 | - | - |

Cldn3
Genomic Map