Assay Details
| Gene Symbol: Dpysl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009955.3 | NP_034085.2 | 11 - 12 | 1510 | 65 | |
| GenBank mRNA | BC062955.1 | - | 11 - 12 | 1509 | 30535984 | 65 |
| X87242.1 | - | 11 - 12 | 1407 | 65 | ||
| Y10339.1 | - | 11 - 12 | 1300 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dpysl2
Genomic Map