Assay Details
| Gene Symbol: Dpysl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009955.3 | NP_034085.2 | 12 - 13 | 1697 | 83 | |
| GenBank mRNA | BC062955.1 | - | 12 - 13 | 1696 | 30535984 | 83 |
| X87242.1 | - | 12 - 13 | 1594 | 83 | ||
| Y10339.1 | - | 12 - 13 | 1487 | 83 | ||
More Information
Set Membership: |
|

Dpysl2
Genomic Map