Assay Details
| Gene Symbol: Ctsl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF121837.1 | - | 3 - 4 | 297 | 77 | |
| AF121838.1 | - | 3 - 4 | 297 | 77 | ||
| AF121839.1 | - | 3 - 4 | 297 | 77 | ||
| AK159511.1 | - | 3 - 4 | 334 | 77 | ||
| AK160100.1 | - | 3 - 4 | 334 | 77 | ||
| AK169712.1 | - | 3 - 4 | 334 | 77 | ||
| AK190042.1 | - | 2 - 3 | 270 | 77 | ||
| J02583.1 | - | 3 - 4 | 310 | 77 | ||
| X06086.1 | - | 3 - 4 | 319 | 77 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ctsl
Genomic Map