Assay Details
| Gene Symbol: Clip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039162.2 | NP_001034251.1 | 13 - 14 | 3230 | 94 | |
| NM_009990.3 | NP_034120.2 | 14 - 15 | 3335 | 94 | ||
| GenBank mRNA | AK045967.1 | - | 14 - 15 | 3180 | 94 | |
| AK172928.1 | - | 14 - 15 | 3080 | 94 | ||
| BC039162.1 | - | 13 - 14 | 3170 | 3498067 | 94 | |
| BC053048.1 | - | 13 - 14 | 3230 | 5709279 | 94 | |
| BC121191.1 | - | 13 - 14 | 2874 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Clip2
Genomic Map