Assay Details
| Gene Symbol: Dazl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277863.1 | NP_001264792.1 | 3 - 4 | 460 | 121 | |
| NM_010021.5 | NP_034151.3 | 3 - 4 | 460 | 121 | ||
| GenBank mRNA | AK014854.1 | - | 3 - 4 | 440 | 121 | |
| AK033183.1 | - | 3 - 4 | 440 | 121 | ||
| AK033316.1 | - | 3 - 4 | 440 | 121 | ||
| AK160855.1 | - | 3 - 4 | 319 | 121 | ||
| BC099940.1 | - | 3 - 4 | 420 | 30048233 | 121 | |
| U38690.1 | - | 3 - 4 | 575 | 121 | ||
| U46694.1 | - | 3 - 4 | 356 | 121 | ||
| X95724.1 | - | 3 - 4 | 349 | 121 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dazl
Genomic Map