Assay Details
| Gene Symbol: Dgat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010046.2 | NP_034176.1 | 8 - 9 | 817 | 103 | |
| GenBank mRNA | AB057816.1 | - | 8 - 9 | 805 | 103 | |
| AF078752.1 | - | 8 - 9 | 875 | 103 | ||
| AK215091.1 | - | 2 - 3 | 99 | 103 | ||
| BC003717.1 | - | 8 - 9 | 817 | 3486724 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dgat1
Genomic Map