Assay Details
| Gene Symbol: Dgat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010046.2 | NP_034176.1 | 13 - 14 | 1161 | 62 | |
| GenBank mRNA | AB057816.1 | - | 13 - 14 | 1149 | 62 | |
| AF078752.1 | - | 13 - 14 | 1219 | 62 | ||
| BC003717.1 | - | 13 - 14 | 1161 | 3486724 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dgat1
Genomic Map