Assay Details
| Gene Symbol: Dspp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010080.2 | NP_034210.2 | 3 - 4 | 218 | 67 | |
| GenBank mRNA | BC116172.1 | - | 3 - 4 | 218 | 67 | |
| BC116173.1 | - | 218 | 67 | |||
| BC129802.1 | - | 218 | 40049023 | 67 | ||
| U67916.1 | - | 3 - 4 | 227 | 67 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2019590 | U67916.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2019590 | NM_010080.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2166132 | BC129802.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2166132 | U67916.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2166132 | BC116173.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2166132 | NM_010080.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 738313 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 578983 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 318171 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 782053 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 659191 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 12940 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P106160 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P175073 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2019590 | U67916.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2019590 | NM_010080.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2166132 | BC129802.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2166132 | U67916.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2166132 | BC116173.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2166132 | NM_010080.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P395991 | U67916.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P395991 | NM_010080.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P175073 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P106160 | - | - |
| Whole Mouse Genome G4122A | A_51_P395991 | U67916.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P395991 | NM_010080.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116932 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125294 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P175073 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P106160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23950747 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04748021 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04762916 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23950748 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23950824 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23950866 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04783229 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23950822 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04760826 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04762909 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04469346 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23950739 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23950779 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23950820 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23950805 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04762907 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23950757 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23950772 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23950865 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23950786 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23950827 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04727862 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04447111 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04762903 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04469360 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23950784 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23950745 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23950761 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04469341 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23950803 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04727867 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23950758 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04162987 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23950783 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23950738 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04748014 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23950767 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04762915 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23950752 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23950774 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23950780 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23950828 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23950794 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23950817 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23950764 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23950863 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23950756 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04986631 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04715800 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23950754 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04762895 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04727870 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04748016 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23950778 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04748020 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04191185 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04762898 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23950791 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23950776 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23950802 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23950775 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23950806 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04727864 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23950782 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23950740 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10523687 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10523685 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10523684 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10523688 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10523686 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10523689 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10523690 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10523691 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10523692 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422246_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10523683 | - | - |
| Mouse Genome 430 2.0 Array | 1422246_at | - | - |
| Mouse Genome 430 2.0 Array | 1422246_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 666279_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 666279_PM_at | - | - |
| Mouse Expression Set 430 | 1422246_PM_at | - | - |
| Mouse Expression Set 430 | 1422246_at | - | - |
| Murine Genome U74v2 Set | 94721_at | - | - |
| Murine Genome U74v2 Set | 94721_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422246_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422246_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422246_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422246_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 864538 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5890403 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5890403 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005890403 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1234325 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1234325 | - | - |
| mouse-6 v1.1 (Illumina) | scl013517.10_7-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5890403 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl013517.10_7-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5890403 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1234325 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1234325 | - | - |

Dspp
Genomic Map