Assay Details
| Gene Symbol: Fhl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289533.1 | NP_001276462.1 | 2 - 3 | 292 | 69 | |
| NM_010212.4 | NP_034342.1 | 2 - 3 | 441 | 69 | ||
| GenBank mRNA | AF055889.1 | - | 1 - 2 | 69 | ||
| AF114381.1 | - | 1 - 2 | 69 | |||
| AF153340.1 | - | 1 - 2 | 213 | 69 | ||
| AK052936.1 | - | 2 - 3 | 293 | 69 | ||
| AK133708.1 | - | 2 - 3 | 414 | 69 | ||
| AK147024.1 | - | 2 - 3 | 442 | 69 | ||
| BC021468.1 | - | 2 - 3 | 350 | 5066565 | 69 | |
| U77040.2 | - | 2 - 3 | 331 | 69 | ||
More Information
Set Membership: |
|

Fhl2
Genomic Map