Assay Details
| Gene Symbol: Gpd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010271.2 | NP_034401.1 | 3 - 4 | 376 | 104 | |
| GenBank mRNA | AK004565.1 | - | 3 - 4 | 381 | 104 | |
| AK018733.1 | - | 3 - 4 | 382 | 104 | ||
| AK137134.1 | - | 3 - 4 | 383 | 104 | ||
| AK220445.1 | - | 3 - 4 | 370 | 104 | ||
| BC005756.1 | - | 3 - 4 | 343 | 104 | ||
| BC019391.1 | - | 3 - 4 | 376 | 4194849 | 104 | |
| J02655.1 | - | 3 - 4 | 368 | 104 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gpd1
Genomic Map