Assay Details
| Gene Symbol: Il18rap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010553.4 | NP_034683.1 | 3 - 4 | 337 | 92 | |
| GenBank mRNA | AF077347.1 | - | 3 - 4 | 346 | 92 | |
| AK053176.1 | - | 3 - 4 | 252 | 92 | ||
| AK137541.1 | - | 3 - 4 | 298 | 92 | ||
| BC120598.1 | - | 3 - 4 | 344 | 40129521 | 92 | |
| BC120600.1 | - | 3 - 4 | 344 | 40129523 | 92 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF077347.1,AK053176.1,AK137541.1,BC120598.1,BC120600.1,NM_010553.4 transcripts.
Set Membership: |
|

Il18rap
Genomic Map