Assay Details
| Gene Symbol: Il18rap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010553.4 | NP_034683.1 | 4 - 5 | 493 | 84 | |
| GenBank mRNA | AF077347.1 | - | 4 - 5 | 502 | 84 | |
| AK053176.1 | - | 4 - 5 | 408 | 84 | ||
| AK137541.1 | - | 4 - 5 | 454 | 84 | ||
| BC120598.1 | - | 4 - 5 | 500 | 40129521 | 84 | |
| BC120600.1 | - | 4 - 5 | 500 | 40129523 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Il18rap
Genomic Map