| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0800021011.mm.1 |
NM_001145831.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800021011.mm.1 |
NM_001145831.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800021011.mm.1 |
NM_001145832.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800021011.mm.1 |
NM_001145832.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800021011.mm.1 |
NM_010631.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800021011.mm.1 |
NM_010631.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800021011.mm.1 |
TC0800002739.mm.1 |
MTA TC |