Assay Details
| Gene Symbol: Kifc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145831.1 | NP_001139303.1 | 15 - 16 | 2031 | 91 | |
| NM_001145832.1 | NP_001139304.1 | 11 - 12 | 1545 | 91 | ||
| NM_010631.2 | NP_034761.3 | 13 - 14 | 1888 | 91 | ||
| GenBank mRNA | AB001457.1 | - | 2 - 3 | 177 | 91 | |
| AF013118.2 | - | 9 - 10 | 1524 | 91 | ||
| AK132895.1 | - | 11 - 12 | 1545 | 91 | ||
| AK135926.1 | - | 8 - 9 | 1087 | 91 | ||
| AK166482.1 | - | 15 - 16 | 91 | |||
| AK203118.1 | - | 1 - 2 | 114 | 92 | ||
| BC004069.1 | - | 5 - 6 | 91 | |||
| BC016118.1 | - | 8 - 9 | 1092 | 91 | ||
| BC023374.1 | - | 6 - 7 | 680 | 91 | ||
| BC070429.1 | - | 13 - 14 | 1888 | 6810893 | 91 | |
| D49546.1 | - | 13 - 14 | 1903 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kifc3
Genomic Map