Assay Details
| Gene Symbol: Capn10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011796.2 | NP_035926.2 | 2 - 3 | 738 | 85 | |
| GenBank mRNA | AF089089.1 | - | 2 - 3 | 770 | 85 | |
| AF126867.1 | - | 2 - 3 | 296 | 85 | ||
| AK002548.1 | - | 2 - 3 | 370 | 85 | ||
| AK013497.1 | - | 2 - 3 | 381 | 85 | ||
| AK088207.1 | - | 2 - 3 | 372 | 85 | ||
| BC005681.1 | - | 2 - 3 | 382 | 3154780 | 85 | |
| BC010969.1 | - | 2 - 3 | 382 | 4159612 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Capn10
Genomic Map