Assay Details
| Gene Symbol: Capn10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011796.2 | NP_035926.2 | 4 - 5 | 1158 | 70 | |
| GenBank mRNA | AF089089.1 | - | 4 - 5 | 1190 | 70 | |
| AF126867.1 | - | 4 - 5 | 716 | 70 | ||
| AK088207.1 | - | 4 - 5 | 792 | 70 | ||
| AK184224.1 | - | 1 - 2 | 137 | 70 | ||
| BC005681.1 | - | 4 - 5 | 802 | 3154780 | 70 | |
| BC010969.1 | - | 4 - 5 | 802 | 4159612 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Capn10
Genomic Map