Assay Details
| Gene Symbol: Farsb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278075.1 | NP_001265004.1 | 9 - 10 | 903 | 140 | |
| NM_011811.4 | NP_035941.2 | 9 - 10 | 903 | 140 | ||
| NR_102757.1 | 9 - 10 | 805 | 140 | |||
| NR_102758.1 | 8 - 9 | 847 | 140 | |||
| GenBank mRNA | AK010271.1 | - | 9 - 10 | 866 | 140 | |
| AK012154.1 | - | 9 - 10 | 866 | 140 | ||
| AK045432.1 | - | 9 - 10 | 855 | 140 | ||
| AK076607.1 | - | 9 - 10 | 865 | 140 | ||
| AK134799.1 | - | 8 - 9 | 809 | 140 | ||
| AK167945.1 | - | 9 - 10 | 866 | 140 | ||
| AK168923.1 | - | 9 - 10 | 847 | 140 | ||
| AK169621.1 | - | 8 - 9 | 810 | 140 | ||
More Information
Set Membership: |
|

Farsb
Genomic Map