Assay Details
| Gene Symbol: Ggt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011820.4 | NP_035950.2 | 5 - 6 | 1083 | 59 | |
| GenBank mRNA | AF077765.1 | - | 5 - 6 | 1068 | 59 | |
| AK049084.1 | - | 5 - 6 | 1086 | 59 | ||
| AK052183.1 | - | 5 - 6 | 1084 | 59 | ||
| AK140308.1 | - | 4 - 5 | 1604 | 59 | ||
| AK170326.1 | - | 5 - 6 | 947 | 59 | ||
| BC113775.1 | - | 5 - 6 | 807 | 40059928 | 59 | |
More Information
Set Membership: |
|

Ggt5
Genomic Map