Assay Details
| Gene Symbol: Ggt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011820.4 | NP_035950.2 | 9 - 10 | 1666 | 80 | |
| GenBank mRNA | AF077765.1 | - | 9 - 10 | 1651 | 80 | |
| AK029138.1 | - | 2 - 3 | 202 | 80 | ||
| AK049084.1 | - | 9 - 10 | 1670 | 80 | ||
| AK052183.1 | - | 9 - 10 | 1667 | 80 | ||
| AK140308.1 | - | 8 - 9 | 2187 | 80 | ||
| AK170326.1 | - | 9 - 10 | 1530 | 80 | ||
| BC089585.1 | - | 5 - 6 | 1085 | 80 | ||
| BC113775.1 | - | 9 - 10 | 1390 | 40059928 | 80 | |
More Information
Set Membership: |
|

Ggt5
Genomic Map