Assay Details
| Gene Symbol: Exo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012012.4 | NP_036142.2 | 5 - 6 | 572 | 58 | |
| GenBank mRNA | AJ238213.1 | - | 3 - 4 | 427 | 58 | |
| AK028728.1 | - | 5 - 6 | 553 | 58 | ||
| AK166425.1 | - | 5 - 6 | 548 | 58 | ||
| BC006671.1 | - | 5 - 6 | 536 | 3499406 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Exo1
Genomic Map