Assay Details
| Gene Symbol: Exo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012012.4 | NP_036142.2 | 10 - 11 | 1425 | 67 | |
| GenBank mRNA | AJ238213.1 | - | 8 - 9 | 1280 | 67 | |
| AK028728.1 | - | 10 - 11 | 1406 | 67 | ||
| AK166425.1 | - | 10 - 11 | 1401 | 67 | ||
| BC006671.1 | - | 10 - 11 | 1389 | 3499406 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Exo1
Genomic Map