Assay Details
| Gene Symbol: Ctbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198859.1 | NP_001185788.1 | 3 - 4 | 525 | 69 | |
| NM_001198860.1 | NP_001185789.1 | 4 - 5 | 718 | 69 | ||
| NM_001198861.1 | NP_001185790.1 | 4 - 5 | 718 | 69 | ||
| NM_013502.3 | NP_038530.1 | 3 - 4 | 525 | 69 | ||
| GenBank mRNA | AB033122.1 | - | 3 - 4 | 490 | 69 | |
| AJ010483.1 | - | 3 - 4 | 438 | 69 | ||
| AK133816.1 | - | 4 - 5 | 689 | 69 | ||
| AK149906.1 | - | 4 - 5 | 696 | 69 | ||
| AK160658.1 | - | 4 - 5 | 708 | 69 | ||
| AK165276.1 | - | 4 - 5 | 673 | 69 | ||
| AK170133.1 | - | 2 - 3 | 232 | 69 | ||
| AK171650.1 | - | 2 - 3 | 358 | 69 | ||
| AK218288.1 | - | 2 - 3 | 237 | 69 | ||
| BC013702.1 | - | 3 - 4 | 438 | 3155102 | 69 | |
| BC015071.1 | - | 3 - 4 | 494 | 4192052 | 69 | |
| BC042425.1 | - | 4 - 5 | 699 | 5005681 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ctbp1
Genomic Map