Assay Details
| Gene Symbol: Gnb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013530.1 | NP_038558.1 | 4 - 5 | 588 | 62 | |
| GenBank mRNA | AK014789.1 | - | 4 - 5 | 307 | 62 | |
| AK030354.1 | - | 3 - 4 | 529 | 62 | ||
| BC018239.1 | - | 4 - 5 | 588 | 4511683 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gnb3
Genomic Map